Läs mer
Retorphan, TRC
Produktkod. 30423043
Change view
Kvantitet:
1 mg
5 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30423043 50 mg
30412925 1 mg
30412360 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30423043 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR27850050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 112573-72-5
Molekylformel C21 H23 N O4 S
Molekylvikt (g/mol) 385.48
Synonym N-[(2R)-2-[(Acetylthio)methyl]-1-oxo-3-phenylpropyl]glycine Phenylmethyl Ester, Dexecadotril
IUPAC-namn benzyl 2-[[(2R)-2-(acetylsulfanylmethyl)-3-phenylpropanoyl]amino]acetate
LEDER CC(=O)SC[C@H](Cc1ccccc1)C(=O)NCC(=O)OCc2ccccc2
Specifikationer
Kemiskt namn eller material Retorphan
Kvantitet 50 mg
InChI formel InChI=1S/C21H23NO4S/c1-16(23)27-15-19(12-17-8-4-2-5-9-17)21(25)22-13-20(24)26-14-18-10-6-3-7-11-18/h2-11,19H,12-15H2,1H3,(H,22,25)/t19-/m0/s1
Formel vikt 385.1348