Läs mer
Brady™ Retractable Barrier Belt with Chrome Posts
Produktkod. 11791126
:
Length: 3.5m; Color: Green
Produktkod. 11791126
Brand: Brady™ 912651
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Specifikationer
Produkttyp Retractable Barrier Belt
Färg Green
Material Chrome
Höjd (metrisk) 100 mm
Inkluderar Retractable Barrier Belt - 3.5 Metres with Chrome Posts
Längd (metrisk) 3.5 m
Kvantitet 1 Ea.
Vikt (metrisk) 12 kg