Läs mer
Revizinone, MedChemExpress
Produktkod. 16420679
Encompass_Preferred
Change view
Kvantitet:
1 mg
5 mg
10 mg
50 mg
100 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16420679 1 mg
16489656 50 mg
16484396 100 mg
16445837 5 mg
16413108 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16420679 Leverantör Medchem Express Leverantörsnummer HY100615
MedChemExpress Revizinone is a novel selective phosphodiesterase (PDE) inhibitor with IC50 values on this enzyme to 0.036 microM.
Revizinone is a novel selective phosphodiesterase (PDE) inhibitor with IC50 values on this enzyme to 0.036 microM.
Specifikationer
Kemiskt namn eller material Revizinone
Fysisk form Solid
CAS 133718-29-3
Färg White
Kvantitet 1 mg
Anteckningar om renhetsgrad Research
Molekylformel C26H29N5O3
För användning med (applikation) COVID-19-immunoregulation
Synonym R80122
Löslighetsinformation DMSO : ≥ 4.6 mg/mL (10.01 mM)
LEDER O=C(N(C1CCCCC1)C)CO/N=C(C2=CC=CC=C2)/C3=CC4=C(N=C(NC(C5)=O)N5C4)C=C3.[(E)]
Molekylvikt (g/mol) 459.54
Formel vikt 459.54
Procent renhet 98.31%