missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ Rex1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA581488
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Mouse ZFP42.
Involved in the reprogramming of X-chromosome inactivation during the acquisition of pluripotency. Required for efficient elongation of TSIX, a non-coding RNA antisense to XIST. Binds DXPas34 enhancer within the TSIX promoter. Involved in ES cell self-renewal. [UniProt].
Specifikationer
| Rex1 | |
| Polyclonal | |
| PBS with no preservative | |
| P22227 | |
| ZFP42 | |
| A synthetic peptide corresponding to the N-terminus of the Mouse ZFP42. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| ZFP42 | |
| 1700021P10Rik; 2610511M11Rik; AW050347; ELOABP1; EloA-BP1; elongin A binding protein 1; elongin A-binding protein 1; elongin-A-binding protein 1; hREX-1; KIAA1138; mREX-1; reduced expression protein 1; REX1; Rex-1; REX1 transcription factor; REX1, RNA exonuclease 1 homolog; REX1, RNA exonuclease 1 homolog (S. cerevisiae); REXO1; RNA exonuclease 1 homolog; Tceb3bp1; transcription elongation factor B polypeptide 3 binding protein 1; transcription elongation factor B polypeptide 3-binding protein 1; Zfp42; Zfp-42; ZFP42 zinc finger protein; Zfp971; Zinc finger protein 42; zinc finger protein 42 homolog; Zinc finger protein 754; zinc finger protein 971; ZNF754 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 22702 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering