Läs mer
RF9, MedChemExpress
Produktkod. 16466817
Encompass_Preferred
Change view
Kvantitet:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16466817 5 mg
16461079 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16466817 Leverantör Medchem Express Leverantörsnummer HY107382
MedChemExpress RF9 is a potent and selective Neuropeptide FF receptor antagonist, with Ki values of 58 and 75 nM for hNPFF1R and hNPFF2R, respectively.
RF9 is a potent and selective Neuropeptide FF receptor antagonist, with Ki values of 58 and 75 nM for hNPFF1R and hNPFF2R, respectively.
Specifikationer
Kemiskt namn eller material RF9
Fysisk form Solid
CAS 876310-60-0
Färg White
Kvantitet 5 mg
Anteckningar om renhetsgrad Research
Molekylformel C26H38N6O3
För användning med (applikation) COVID-19-immunoregulation
Löslighetsinformation DMSO : 125 mg/mL (259.00 mM; Need ultrasonic) ∣H2O : 16.67 mg/mL (34.54 mM; Need ultrasonic)
LEDER NC([C@H](CC1=CC=CC=C1)NC([C@H](CCCNC(N)=N)NC(C23C[C@H](C4)C[C@H](C[C@H]4C3)C2)=O)=O)=O
Molekylvikt (g/mol) 482.62
Formel vikt 482.62
Procent renhet 98.66%