Learn More
RFPL1/2/3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13215-1-AP-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16881324
  • 1236.20 SEK / 20µL
Estimated Shipment: 08-12-2021
Specifications
RFPL1/2/3
0.22 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O75677, O75678, O75679
ret finger protein like 3, RFPL3
Rabbit
IgG
20 μL
-20°C
Primary
10738, 10739, 5988
Western Blot
Unconjugated
RFPL2
Liquid
RFPL1, RFPL2, RFPL3
RFPL1/2/3 Fusion Protein Ag4090
Antigen Affinity Chromatography