missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFPL1/2/3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | RFPL1/2/3 |
---|---|
Concentration | 0.22 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16881324
|
Proteintech
13215-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 03-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16871324
|
Proteintech
13215-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 03-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
RFPL1/2/3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
O75677, O75678, O75679 | |
10738, 10739, 5988 | |
RFPL1/2/3 Fusion Protein Ag4090 | |
Primary | |
-20°C | |
RFPL2 |
0.22 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ret finger protein like 3, RFPL3 | |
RFPL1, RFPL2, RFPL3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |