missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Description
RFTN2 Polyclonal antibody specifically detects RFTN2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | RFTN2 |
| Användningsområden | Western Blot |
| Klassificering | Polyclonal |
| Konjugera | Unconjugated |
| Utspädning | Western Blot 1:500-1:2000 |
| Formulering | PBS (pH 7.3), 50% glycerol |
| Gene Alias | C2orf11, chromosome 2 open reading frame 11, FLJ30574, MGC117313, raftlin family member 2, raftlin-2, Raft-linking protein 2 |
| Värd art | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 412-501 of human RFTN2 (NP_653230.2). KKASRHIKGEDKNKATSRSIGLDTTSSQPAESRHLPEECRLSPSRECWTKEGRLAQHNSFSGFSSSDNVLRELDDGQFDQEDGVTQVTCM |
| Reningsmetod | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?