missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFX3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14784-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the regulatory factor X gene family, which encodes transcription factors that contain a highly-conserved winged helix DNA binding domain. The protein encoded by this gene is structurally related to regulatory factors X1, X2, X4, and X5. It is a transcriptional activator that can bind DNA as a monomer or as a heterodimer with other RFX family members. Two transcript variants encoding different isoforms have been described for this gene, and at least one of the variants utilizes alternative polyadenylation signals.Specifications
RFX3 | |
Polyclonal | |
Unconjugated | |
RFX3 | |
Regulatory factor X 3, RFX3, Transcription factor RFX3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19726, 361746, 5991 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P48380, P48381 | |
Rfx3 | |
RFX3 Fusion Protein Ag6413 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |