missing translation for 'onlineSavingsMsg'
Learn More

RGS9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 17970-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16877984

  • 1236.20 SEK / 20µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody got about 58kD in the mouse eye and 70kD in the mouse brain in western blotting, both in line with their expected masses. Defects in RGS9 are a cause of prolonged electroretinal response suppression (PERRS).

The cascade of biochemical reactions within the vision system is complex and contains numerous steps taking place in oseconds. RGS is a family of GTPase accelerating proteins (GAPs). RGS9 is a member of this family and is the predomit source of GAP activity in rod outer segments. RGS9-1, found predominately in vertebrate cone and rod photoreceptors, is a useful model for understanding how rapid recovery kinetics of the light responses in retinal photoreceptors take place.
Specifications

Specifications

RGS9
0.37 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O54828, O75916, P49805
PERRS, RGS9, RGS9L
Rabbit
IgG
20 μL
-20°C
Primary
19739, 29481, 8787
Western Blot
Unconjugated
RGS9
Liquid
RGS9
RGS9 Fusion Protein Ag11976
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers