Läs mer
RH 1788, TRC
Produktkod. 30415201
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30415201 250 mg
30426984 100 mg
30418870 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30415201 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR315765250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 163860-36-4
Molekylformel C22 H28 N2 O3
Molekylvikt (g/mol) 368.4693
Synonym Benzoic acid, 3,5-dimethyl-, 1-(1,1-dimethylethyl)-2-[4-(1-hydroxyethyl)benzoyl]hydrazide, RH 1788
IUPAC-namn N-tert-butyl-N'-[4-(1-hydroxyethyl)benzoyl]-3,5-dimethylbenzohydrazide
LEDER CC(O)c1ccc(cc1)C(=O)NN(C(=O)c2cc(C)cc(C)c2)C(C)(C)C
Specifikationer
Kemiskt namn eller material Tebufenozide-1-hydroxyethyl
Kvantitet 250 mg
InChI formel InChI=1S/C22H28N2O3/c1-14-11-15(2)13-19(12-14)21(27)24(22(4,5)6)23-20(26)18-9-7-17(8-10-18)16(3)25/h7-13,16,25H,1-6H3,(H,23,26)
Formel vikt 368.21