Läs mer
RH 9886, TRC
Produktkod. 30418707
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30418707 10 mg
30417762 5 mg
30412037 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30418707 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR31577010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 166547-61-1
Molekylformel C22 H28 N2 O3
Molekylvikt (g/mol) 368.47
Synonym Benzoic acid, 3-(hydroxymethyl)-5-methyl-, 1-(1,1-dimethylethyl)-2-(4-ethylbenzoyl)hydrazide, RH 9886
IUPAC-namn N-tert-butyl-N'-(4-ethylbenzoyl)-3-(hydroxymethyl)-5-methylbenzohydrazide
LEDER CCc1ccc(cc1)C(=O)NN(C(=O)c2cc(C)cc(CO)c2)C(C)(C)C
Specifikationer
Kemiskt namn eller material Tebufenozide-hydroxymethyl
Kvantitet 10 mg
InChI formel InChI=1S/C22H28N2O3/c1-6-16-7-9-18(10-8-16)20(26)23-24(22(3,4)5)21(27)19-12-15(2)11-17(13-19)14-25/h7-13,25H,6,14H2,1-5H3,(H,23,26)
Formel vikt 368.21