missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHAMM/Lgals9 Rabbit anti-Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15819-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This antibody is specific for rat HAMM, not react with Human and Mouse HAMM.
The protein encoded by this gene is involved in cell motility. It is expressed in breast tissue and together with other proteins, it forms a complex with BRCA1 and BRCA2, thus is potentially associated with higher risk of breast cancer. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.Specifications
RHAMM/Lgals9 | |
Polyclonal | |
Unconjugated | |
Hmmr | |
Hmmr, RHAMM | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15366, 25460 | |
-20°C | |
Liquid |
Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P97779, Q00547 | |
Hmmr | |
Peptide | |
150 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |