Learn More
BD Rhapsody™ Mouse TCR/BCR Next Amplification Kit
Product Code. 32226998 Shop All BD Biosciences Products
Change view
Click to view available options
Product Type:
Mouse TCR/BCR Next Amplification Kit
Unit Size:
1 test
Product Code. Product Type Unit Size
32226998 Mouse TCR/BCR Next Amplification Kit 1 test
1 options
Product Code. 32226998 Supplier BD Supplier No. 667059
 Restricted Item
Beräknad leverans: 18-11-2026
The next generation of BD Rhapsody™ TCR/BCR Multiomic Assays reveal immune repertoire richness with great sensitivity alongside simultaneous single-cell gene and protein expression analyses.
Validated on our new Enhanced Cell Capture Beads V3, BD Rhapsody™ TCR/BCR Next Multiomic Assays enable a highly sensitive analysis of full-length TCR/BCR repertoires with robust workflows for human and mouse. Key features include:
  • Superior full-length TCR and BCR pairing metrics
  • Multiomics-enabled for simultaneous immune profiling alongside protein and gene expression analyses in the same cell
  • Compatibility with preserved cells and BD™ OMICS-Guard Sample Preservation Buffer
  • Intuitive data analysis pipeline that provides unbiased insights about the biology of your samples
Kit provides the necessary reagents to prepare single-cell full-length Mouse TCR and BCR libraries that are compatible with both BD Rhapsody™ Whole Transcriptome Analysis (WTA) and BD Rhapsody™ Targeted mRNA assays.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Mouse TCR/BCR Next Amplification Kit