missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Rhenium(IV) oxide, 99.9% (metals basis)

Brand:  Alfa Aesar™ 046538.06

Additional Details : CAS Number : 12036-09-8 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15452717

  • 3904.80 SEK / 5g


Estimated Shipment date
from Supplier 27-08-2018
Add to basket

Specifications

Chemical Identifiers

12036-09-8
218.205
WXBOMIKEWRRKBB-UHFFFAOYSA-N
O2Re
MFCD00016305
82847

Specifications

Rhenium (IV) oxide
99.9%
O2Re
MFCD00016305
O=[Re]=O
218.205
218.21
Odorless
5g
12036-09-8
(metals basis)
ReO2
WXBOMIKEWRRKBB-UHFFFAOYSA-N
dioxorhenium
82847
Powder
1000°C (decomposition)
Videos
SDS