missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ Rho Pan Polyclonal Antibody
GREENER_CHOICE

Produktkod. 16683295 Handla allt Thermo Scientific Produkter
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
Förpackningsstorlek:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16683295 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16683295 Leverantör Invitrogen™ Leverantörsnummer PA5106862

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RHOC.

This gene encodes a member of the Rho family of small GTPases, which cycle between inactive GDP-bound and active GTP-bound states and function as molecular switches in signal transduction cascades. Rho proteins promote reorganization of the actin cytoskeleton and regulate cell shape, attachment, and motility. The protein encoded by this gene is prenylated at its C-terminus, and localizes to the cytoplasm and plasma membrane. It is thought to be important in cell locomotion. Overexpression of this gene is associated with tumor cell proliferation and metastasis. Multiple alternatively spliced variants, encoding the same protein, have been identified.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Rho Pan
Användningsområden Immunohistochemistry (Paraffin), Western Blot
Klassificering Polyclonal
Koncentration 1 mg/mL
Konjugera Unconjugated
Formulering PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gen RHOC
Genaccessionsnr. P08134, P17081, P61586, P61589, P62745, P62746, P62747, Q62159, Q8BYP3, Q8R527, Q9HBH0, Q9JJL4, Q9QUI0
Gene Alias AA017882; AI324259; AI845056; Aplysia ras-related homolog 12; Aplysia RAS-related homolog 6; aplysia ras-related homolog 9 (RhoC); aplysia ras-related homolog A; aplysia ras-related homolog A1; aplysia ras-related homolog A2; aplysia ras-related homolog B; ARH12; ARH6; ARH9; ARHA; Arha1; Arha2; Arhb; ARHC; ARHF; ARHQ; AV026554; epididymis secretory protein Li 42; h12; h6; H9; HEL-S-42; Ifld1; induced in fatty liver dystrophy 1; MST081; MSTP081; oncogene RHO H12; oncogene RHO H6; oncogene RHO H9; plysia ras-related homolog A2; Ras family member A; ras homolog 9 (RhoC); ras homolog A1; ras homolog A2; ras homolog B; ras homolog family member A; ras homolog family member B; ras homolog family member C; ras homolog family member F (in filopodia); ras homolog family member F, filopodia associated; ras homolog family member Q; ras homolog gene family, member A; ras homolog gene family, member A1; ras homolog gene family, member A2; ras homolog gene family, member AB; ras homolog gene family, member B; ras homolog gene family, member C; ras homolog gene family, member f; ras homolog gene family, member F (in filopodia); ras homolog gene family, member Q; RASL7A; ras-like protein family member 7A; Ras-like protein TC10; RAS-like, family 7, member A; ras-related GTP-binding protein TC10; RAS-related homolog 9; Rho cDNA clone 12; Rho cDNA clone 6; rho cDNA clone 9; rho family; Rho family GTPase; rho family GTPase Rif; Rho in filopodia; RHO12; Rhoa; Rhob; RHOC; rhoC GTPase; RHOF; RHOH12; RHOH6; RHOH9; Rhoq; rho-related GTP-binding protein RhoB; Rho-related GTP-binding protein RhoC; rho-related GTP-binding protein RhoF; Rho-related GTP-binding protein RhoQ; RIF; SIG-61; Silica-induced gene 61 protein; small GTP binding protein RhoA; small GTP binding protein RhoC; small GTPase Tc10; TC10; TC10A; Transforming protein RhoA
Gensymboler RHOA, RHOB, RHOC, RHOF, RHOQ
Värd art Rabbit
Immunogen A synthesized peptide derived from human RHOC(Accession P08134), corresponding to amino acid residues K162-L193.
Reningsmetod Affinity chromatography
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 104215, 117273, 11848, 11852, 11853, 23433, 23912, 295342, 387, 388, 389, 54509, 64373, 690130, 85428
Målarter Human, Mouse, Rat
Innehåll och lagring -20°C
Produkttyp Antibody
Form Liquid
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.