Läs mer
Rhynchophylline, TRC
Produktkod. 30138102
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138102 100 mg
30132681 25 mg
30003548 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138102 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR414180100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 76-66-4
Molekylformel C22 H28 N2 O4
Molekylvikt (g/mol) 384.47
Synonym Spiro[3H-indole-3,1'(5'H)-indolizine]-7'-acetic acid, 6'-ethyl-1,2,2',3',6',7',8',8'a-octahydro-α-(methoxymethylene)-2-oxo-, methyl ester, (αE,1'R,6'R,7'S,8'aS)-, Corynoxan-16-carboxylic acid, 16,17-didehydro-17-methoxy-2-oxo-, methyl ester, (7β,16E,20α)-, Rhyncophylline (6CI,7CI,8CI), Mitrinermin, Mitrinermine, NSC 21731, Rhynchophyllin, Rhynchophylline, Spiro[3H-indole-3,1'(5'H)-indolizine]-7'-acetic acid, 6'-ethyl-1,2,2',3',6',7',8',8'a-octahydro-α-(methoxymethylene)-2-oxo-, methyl ester, [1'R-[1'α,6'β,7'α(E),8'aβ]]-
IUPAC-namn methyl (E)-2-[(3R,6'R,7'S,8'aS)-6'-ethyl-2-oxospiro[1H-indole-3,1'-3,5,6,7,8,8a-hexahydro-2H-indolizine]-7'-yl]-3-methoxyprop-2-enoate
LEDER CC[C@H]1CN2CC[C@@]3([C@@H]2C[C@@H]1\C(=C/OC)\C(=O)OC)C(=O)Nc4ccccc34
Specifikationer
Kemiskt namn eller material Rhynchophylline
Kvantitet 100 mg
InChI formel InChI=1S/C22H28N2O4/c1-4-14-12-24-10-9-22(17-7-5-6-8-18(17)23-21(22)26)19(24)11-15(14)16(13-27-2)20(25)28-3/h5-8,13-15,19H,4,9-12H2,1-3H3,(H,23,26)/b16-13+/t14-,15-,19-,22+/m0/s1
Formel vikt 384.2049