Läs mer
Ribavirin-13C5, TRC
Produktkod. 30004877
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30004877 10 mg
30004844 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30004877 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR41447710MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1646818-35-0
Molekylformel 13C5 C3 H12 N4 O5
Molekylvikt (g/mol) 249.17
Synonym Ribavirin 13C5, 1H-1,2,4-Triazole-3-carboxamide, 1-β-D-ribofuranosyl-1,2,3,4,5-13C5-
IUPAC-namn 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxy(1^{13}C)methyl)(2,3,4,5-^{13}C_{4})oxolan-2-yl]-1,2,4-triazole-3-carboxamide
LEDER NC(=O)c1ncn(n1)[13C@@H]2O[13C@H]([13CH2]O)[13C@@H](O)[13C@H]2O
Specifikationer
Kemiskt namn eller material Ribavirin 13C5
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C8H12N4O5/c9-6(16)7-10-2-12(11-7)8-5(15)4(14)3(1-13)17-8/h2-5,8,13-15H,1H2,(H2,9,16)/t3-,4-,5-,8-/m1/s1/i1+1,3+1,4+1,5+1,8+1
Formel vikt 249.0975
Procent renhet >95%