Läs mer
Ribociclib succinate hydrate, MedChemExpress
Produktkod. 16499367
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16499367 5 mg
16472897 50 mg
16456828 10 mg
16456936 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16499367 Leverantör Medchem Express Leverantörsnummer HY15777C
MedChemExpress Ribociclib succinate hydrate (LEE011 succinate hydrate) is a highly specific CDK4/6 inhibitor with IC50 values of 10 nM and 39 nM, respectively, and is over 1,000-fold less potent against the cyclin B/CDK1 complex.
Ribociclib succinate hydrate (LEE011 succinate hydrate) is a highly specific CDK4/6 inhibitor with IC50 values of 10 nM and 39 nM, respectively, and is over 1,000-fold less potent against the cyclin B/CDK1 complex.
Specifikationer
Kemiskt namn eller material Ribociclib succinate hydrate
Fysisk form Powder
CAS 1374639-79-8
Färg White
Kvantitet 5 mg
Anteckningar om renhetsgrad Research
Molekylformel C23H30N8O.C4H6O4.xH2O
För användning med (applikation) Cancer-Kinase/protease
Synonym LEE011 succinate hydrate
Löslighetsinformation DMSO : 100 mg/mL (175.24 mM; Need ultrasonic) ∣H2O : 4 mg/mL (7.01 mM; Need ultrasonic)
LEDER O=C(N(C)C)C(N1C2CCCC2)=CC(C1=N3)=CN=C3NC(N=C4)=CC=C4N5CCNCC5.OC(CCC(O)=O)=O.[F,Cl,Br,I].O
Molekylvikt (g/mol) 570.64
Formel vikt 570.64
Procent renhet 99.96%