Läs mer
Ribostamycin sulfate, MedChemExpress
Produktkod. 16404577
Encompass_Preferred
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
16404577 50 mg
1 options
Produktkod. 16404577 Leverantör Medchem Express Leverantörsnummer HYB1228
MedChemExpress Ribostamycin sulfate (Vistamycin sulfate) is a broad-spectrum antimicrobial, inhibits bacterial protein synthesis at the level of 30S and 50S ribosomal subunit binding, also inhibits the chaperone activity of protein disulfide isomerase (PDI), used in pharmacokinetic and nephrotoxicity studies
Ribostamycin sulfate (Vistamycin sulfate) is a broad-spectrum antimicrobial, inhibits bacterial protein synthesis at the level of 30S and 50S ribosomal subunit binding, also inhibits the chaperone activity of protein disulfide isomerase (PDI), used in pharmacokinetic and nephrotoxicity studies
Specifikationer
Kemiskt namn eller material Ribostamycin sulfate
Fysisk form Solid
CAS 53797-35-6
Färg White
Kvantitet 50 mg
Anteckningar om renhetsgrad Research
Molekylformel C17H36N4O14S
För användning med (applikation) COVID-19-immunoregulation
Synonym Vistamycin sulfate
Löslighetsinformation H2O : 130 mg/mL (235.27 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
LEDER O=S(O)(O)=O.O[C@@H]([C@H](O)[C@@H](CN)O1)[C@@H](N)[C@@]1([H])O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O)[C@@]2([H])O[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O
Molekylvikt (g/mol) 552.55
Formel vikt 552.55
Procent renhet 98.0%