Beskrivning
Three unique packaging configurations allow for easy distribution and transport.
53 Series Containers
- Easy to transport trays ideal for operating rooms and hospitals
- Clear polystyrene containers have screw-top lids
- Comply with all updated federal and state OSHA requirements
- Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
56 Series Containers
- Patented Epredia™ Bio-Tite™ closure system is dual-threaded, making it virtually impossible to cross-thread and minimizing leakage
- Easy to transport trays
- Comply with all updated federal and state OSHA requirements
- Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
59 Series Containers
- Patented Epredia Bio-Tite closure system is dual-threaded, making it virtually impossible to cross-thread and minimizing leakage
- Stack pack reduces excess packaging
- Comply with all updated federal and state OSHA requirements
- Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
Specifikationer
Specifikationer
| Produkttyp | Specimen Container Label |
| Förslutning ingår | Ja |
| Beskrivning | Formalin Warning Labels, Large |