Läs mer
Epredia™ Richard-Allan Scientific™ Gomori Trichrome (Green Collagen), Gomori Trichrome Stain Solution, Green Collagen
Produktkod. 12533657 Handla allt Epredia Produkter
Klicka för att se tillgängliga alternativ
Typ:
Gomori's Trichrome Stain Solution, Green Collagen
Förpackningsstorlek:
500ml
Produktkod. 12533657
Brand: Epredia™ 88031
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Save time with Epredia™ Richard-Allan Scientific™ Gomori Trichrome (Green Collagen), a one-step trichrome stain that provides distinctive red muscle fibers with green collagen staining.
  • Used to study the diseases of connective tissue and muscle associated with fibrotic and dystrophic changes
TRUSTED_SUSTAINABILITY
Specifikationer
Typ Gomori's Trichrome Stain Solution, Green Collagen