Läs mer
Epredia™ Resolve™ Immersion olja
Handla allt Epredia Produkter
Viskositet:
Hög
Låg
Kvantitet:
0,5 oz.
1 uns
4 oz.
Förpackningsstorlek:
Paket med 12
Beskrivning
- Manufactured using a proprietary process that removes interfering substances and will not damage valuable optics or irreplaceable slide specimens
- Packaged conveniently in four sizes
Product Details:
- Available in both low and high viscosity
- Non-drying and non-hardening
- Refractive index of 1.5150
Specifikationer
Specifikationer
| Typ | Immersion Oil |
| Viskositet | Hög |
| Kvantitet | 1 uns |
| Brytningsindex | 1.515 |