Läs mer
Ridaforolimus, MedChemExpress
Produktkod. 16498138
Encompass_Preferred
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16498138 10 mg
16424057 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16498138 Leverantör Medchem Express Leverantörsnummer HY50908
MedChemExpress Ridaforolimus (MK-8669) is a potent and selective mTOR inhibitor; inhibits ribosomal protein S6 phosphorylation with an IC50 of 0.2 nM in HT-1080 cells.
Ridaforolimus (MK-8669) is a potent and selective mTOR inhibitor
Specifikationer
Kemiskt namn eller material Ridaforolimus
Fysisk form Solid
CAS 572924-54-0
Färg Off-White
Kvantitet 10 mg
Anteckningar om renhetsgrad Research
Molekylformel C53H84NO14P
För användning med (applikation) Cancer-Kinase/protease
Synonym MK-8669 Deforolimus AP23573
Löslighetsinformation DMSO : ≥ 44 mg/mL (44.44 mM)
LEDER O=C([C@@]1(O)[C@@H](CC[C@@H](C[C@@H](/C(C)=C/C=C/C=C/[C@H](C[C@@H](C)C([C@@H]([C@@H](/C(C)=C/[C@H]2C)O)OC)=O)C)OC)O1)C)C(N3CCCC[C@H]3C(O[C@@H](CC2=O)[C@@H](C[C@@H]4C[C@H]([C@H](OP(C)(C)=O)CC4)OC)C)=O)=O
Molekylvikt (g/mol) 990.21
Formel vikt 990.21
Procent renhet 95.0%