Läs mer
Rifampicin N-4’-Oxide, TRC
Produktkod. 30137694
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137694 100 mg
30003475 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137694 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR508020100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 125833-03-6
Molekylformel C43 H58 N4 O13
Molekylvikt (g/mol) 838.94
Synonym Rifamycin, 3-[[(4-methyl-4-oxido-1-piperazinyl)imino]methyl]- (9CI), Rifamycin, 3-[[(4-methyl-1-piperazinyl)imino]methyl]-, N-oxide, 2,7-(Epoxypentadeca[1,11,13]trienimino)naphtho[2,1-b]furan, rifamycin deriv., Rifampicin N-Oxide
LEDER CO[C@H]1\C=C\O[C@@]2(C)Oc3c(C)c(O)c4c(O)c(NC(=O)\C(=C/C=C/[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(=O)C)[C@@H]1C)\C)c(\C=N\N5CC[N+](C)([O-])CC5)c(O)c4c3C2=O
Specifikationer
Kemiskt namn eller material Rifampicin N-4'-Oxide
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C43H58N4O13/c1-21-12-11-13-22(2)42(55)45-33-28(20-44-46-15-17-47(9,56)18-16-46)37(52)30-31(38(33)53)36(51)26(6)40-32(30)41(54)43(8,60-40)58-19-14-29(57-10)23(3)39(59-27(7)48)25(5)35(50)24(4)34(21)49/h11-14,19-21,23-25,29,34-35,39,49-53H,15-18H2,1-10H3,(H,45,55)/b12-11+,19-14+,22-13-,44-20+/t21-,23+,24+,25+,29-,34-,35+,39+,43-/m0/s1
Formel vikt 838.4
Procent renhet >95%