Läs mer
Rifampicin Quinone, TRC
Produktkod. 30417733
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30417733 50 mg
1 options
Produktkod. 30417733 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR50804050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Rifampicin quinone
CAS 13983-13-6
Anteckningar om renhetsgrad HPLC
Molekylformel C43 H56 N4 O12
Kvantitet 50 mg
Synonym Rifamycin','1,4-dideoxy-1,4-dihydro-3-[(E)-[(4-methyl-1-piperazinyl)imino]methyl]-1,4-dioxo-, 2,7-(Epoxypentadeca[1,11,13]trienimino)naphtho[2,1-b]furan-1,6,9,11(2H)-tetrone','5,17,19,21-tetrahydroxy-23-methoxy-2,4,12,16,18,20,22-heptamethyl-8-[N-(4-methyl-1-piperazinyl)formimidoyl]-','21-acetate (8CI), 2,7-(Epoxypentadeca[1,11,13]trienimino)naphtho[2,1-b]furan','rifamycin deriv., NSC 145633, Rifaldazinequinone, Rifampicin quinone, Rifampin quinone
InChI formel InChI=1S/C43H56N4O12/c1-21-12-11-13-22(2)42(55)45-33-28(20-44-47-17-15-46(9)16-18-47)37(52)30-31(38(33)53)36(51)26(6)40-32(30)41(54)43(8,59-40)57-19-14-29(56-10)23(3)39(58-27(7)48)25(5)35(50)24(4)34(21)49/h11-14,19-21,23-25,29,34-35,39,49-51H,15-18H2,1-10H3,(H,45,55)/b12-11+,19-14+,22-13-,44-20+/t21-,23+,24+,25+,29-,34-,35+,39+,43-/m0/s1
LEDER CO[C@H]1\C=C\O[C@@]2(C)Oc3c(C)c(O)c4C(=O)C(=C(\C=N\N5CCN(C)CC5)C(=O)c4c3C2=O)NC(=O)\C(=C/C=C/[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(=O)C)[C@@H]1C)\C
Molekylvikt (g/mol) 820.92
Formel vikt 820.3895