Läs mer
Rifamycin B, TRC
Produktkod. 30010724
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010724 50 mg
30010611 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30010724 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR50817050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13929-35-6
Molekylformel C39 H49 N O14
Molekylvikt (g/mol) 755.8
Synonym Rifamycin, 4-O-(carboxymethyl)-, Acetic acid, [(1,2-dihydro-5,6,17,19,21-pentahydroxy-23-methoxy-2,4,12,16,18,20,22-heptamethyl-1,11-dioxo-2,7-(epoxypentadeca[1,11,13]trienimino)naphtho[2,1-b]furan-9-yl)oxy]-, 21-acetate (7CI,8CI), Rifomycin B (6CI), 2,7-(Epoxypentadeca[1,11,13]trienimino)naphtho[2,1-b]furan, rifamycin deriv., NSC 145604, Nancimycin, Rifamycin B, Rifaximin Imp B (EP)
LEDER CO[C@H]1\C=C\O[C@@]2(C)Oc3c(C)c(O)c4c(O)c(NC(=O)\C(=C/C=C/[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(=O)C)[C@@H]1C)\C)cc(OCC(=O)O)c4c3C2=O
Specifikationer
Kemiskt namn eller material Rifamycin B
Kvantitet 50 mg
InChI formel InChI=1S/C39H49NO14/c1-17-11-10-12-18(2)38(49)40-24-15-26(51-16-27(42)43)28-29(34(24)47)33(46)22(6)36-30(28)37(48)39(8,54-36)52-14-13-25(50-9)19(3)35(53-23(7)41)21(5)32(45)20(4)31(17)44/h10-15,17,19-21,25,31-32,35,44-47H,16H2,1-9H3,(H,40,49)(H,42,43)/b11-10+,14-13+,18-12-/t17-,19+,20+,21+,25-,31-,32+,35+,39-/m0/s1
Formel vikt 755.3153