Läs mer
Rifamycin O, TRC
Produktkod. 30010368
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010368 100 mg
30007217 50 mg
30004306 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010368 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR508190100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 14487-05-9
Molekylformel C39H47NO14
Molekylvikt (g/mol) 753.79
Synonym 4-O-(Carboxymethyl)-1-deoxy-1,4-dihydro-4-hydroxy-1-oxo-rifamycin γ-Lactone, NSC 182391, Rifaximin EP Impurity F;
IUPAC-namn [(7'S,9'E,11'S,12'R,13'S,14'R,15'R,16'R,17'S,18'S,19'E,21'Z)-2',15',17'-trihydroxy-11'-methoxy-3',7',12',14',16',18',22'-heptamethyl-4,6',23',29'-tetraoxo-spiro[1,3-dioxolane-2,27'-8,30-dioxa-24-azatetracyclo[23.3.1.14,7.05,28]triaconta-1,3,5(28),9,19,21,25-heptaene]-13'-yl] acetate
LEDER O=C1C2=C3C(C(C4=CC35OC(CO5)=O)=O)=C(O)C(C)=C2O[C@]1(C)O/C=C/[C@H](OC)[C@@H](C)[C@@H](OC(C)=O)[C@H](C)[C@H](O)[C@H](C)[C@@H](O)[C@@H](C)/C=C/C=C(C)\C(N4)=O
Specifikationer
Kemiskt namn eller material Rifamycin O
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C39H47NO14/c1-17-11-10-12-18(2)37(48)40-24-15-39(51-16-26(42)53-39)29-27(33(24)46)32(45)22(6)35-28(29)36(47)38(8,54-35)50-14-13-25(49-9)19(3)34(52-23(7)41)21(5)31(44)20(4)30(17)43/h10-15,17,19-21,25,30-31,34,43-45H,16H2,1-9H3,(H,40,48)/b11-10+,14-13+,18-12-/t17?,19?,20?,21?,25?,30-,31+,34?,38?,39?/m0/s1
Formel vikt 753.3
Procent renhet >95%