missing translation for 'onlineSavingsMsg'
Learn More

Rifamycin SV sodium salt, MP Biomedicals™

Catalog Number p-4405606
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
14897-39-3
C37H47NNaO12
720.77
MFCD09752825
YVOFSHPIJOYKSH-DITFEXNXSA-N
rifamycin sodium salt, rifamycin sodium usan, rifamycin sv sodium salt, rifocina tn, otofa tn
124219641
sodium (7S,9Z,11S,12R,13S,14R,15R,16R,17S,18S,19Z,21Z)-2,15,17,27,29-pentahydroxy-11-methoxy-3,7,12,14,16,18,22-heptamethyl-6,23-dioxo-8,30-dioxa-24-azatetracyclo[23.3.1.1⁴,⁷.0⁵,²⁸]triaconta-1,3,5(28),9,19,21,25(29),26-octaen-13-yl acetate
[Na+].CO[C@H]1\C=C/O[C@@]2(C)OC3=C(C)C(O)=C4C(O)=C(NC(=O)\C(C)=C/C=C\[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(C)=O)[C@@H]1C)C=C(O)C4=C3C2=O
This item is not returnable. View return policy
14897-39-3
C37H47NNaO12
720.77
MFCD09752825
YVOFSHPIJOYKSH-DITFEXNXSA-N
rifamycin sodium salt, rifamycin sodium usan, rifamycin sv sodium salt, rifocina tn, otofa tn
124219641
sodium (7S,9Z,11S,12R,13S,14R,15R,16R,17S,18S,19Z,21Z)-2,15,17,27,29-pentahydroxy-11-methoxy-3,7,12,14,16,18,22-heptamethyl-6,23-dioxo-8,30-dioxa-24-azatetracyclo[23.3.1.1⁴,⁷.0⁵,²⁸]triaconta-1,3,5(28),9,19,21,25(29),26-octaen-13-yl acetate
[Na+].CO[C@H]1\C=C/O[C@@]2(C)OC3=C(C)C(O)=C4C(O)=C(NC(=O)\C(C)=C/C=C\[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(C)=O)[C@@H]1C)C=C(O)C4=C3C2=O
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'