Läs mer
RIG-1 modulator 1, MedChemExpress
Produktkod. 16491089
Encompass_Preferred
Change view
Kvantitet:
1 mg
5 mg
10 mg
50 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16491089 1 mg
16480507 50 mg
16474158 10 mg
16446867 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16491089 Leverantör Medchem Express Leverantörsnummer HY107902
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
MedChemExpress RIG-1 modulator 1 is an anti-viral compound which can be useful for the treatment of viral infections including influenza virus, HBV, HCV and HIV extracted from patent WO 2015172099 A1.
RIG-1 modulator 1 is an anti-viral compound which can be useful for the treatment of viral infections including influenza virus, HBV, HCV and HIV extracted from patent WO 2015172099 A1.
Specifikationer
Kemiskt namn eller material RIG-1 modulator 1
Fysisk form Solid
CAS 1428729-63-8
Färg Light Yellow
Kvantitet 1 mg
Anteckningar om renhetsgrad Research
Molekylformel C14H17N5OS2
För användning med (applikation) COVID-19-anti-virus
Löslighetsinformation DMSO : ≥ 35 mg/mL (104.34 mM)
LEDER O=C(NC1=NC2=CC=C(SC(C)=N3)C3=C2S1)NCCN(C)C
Molekylvikt (g/mol) 335.45
Formel vikt 335.45
Procent renhet 98.01%