Learn More
Brady™ Rigid Polypropylene Evacuation Panel Safety Signs
98.85 SEK - 181.00 SEK
Specifications
Material | Polypropylene |
---|---|
Quantity | 1 Ea. |
Description | Rigid Polypropylene |
Max. Temperature (Metric) | 120°C |
Color | White on Green |
Product Code | Brand | Displayed Text | Width (Metric) | Height (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Displayed Text | Width (Metric) | Height (Metric) | Price | Quantity & Availability | |||
11725044
|
Brady™
807232 |
Exit and Emergency Exit Down Arrow | 15 mm | 210 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11700843
|
Brady™
807240 |
Exit and Emergency Exit Up Arrow | 10 mm | 210 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11790963
|
Brady™
811158 |
Exit and Emergency Exit Left Arrow | 10 mm | 210 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11720843
|
Brady™
807242 |
Exit and Emergency Exit Up Arrow | 10 mm | 297 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Specifications
Polypropylene | |
Rigid Polypropylene | |
White on Green |
1 Ea. | |
120°C |