Läs mer
Riluzole-13C,15N2, TRC
Produktkod. 30138508
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138508 1 mg
30010717 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30138508 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR5100021MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1215552-03-6
Molekylformel C713CH5F315N2OS
Molekylvikt (g/mol) 237.18
Synonym 6-(Trifluoromethoxy)-2-benzothiazolamine-13C,15N2, 6-(Trifluoromethoxy)-2-amino- benzothiazole-13C,15N2, 2-Amino-6-(trifluoromethoxy)benzothiazole-13C,15N2, PK-26124-13C,15N2, RP-54274-13C,15N2, Rilutek-13C,15N2;
IUPAC-namn 6-(trifluoromethoxy)-1,3-benzothiazol-2-(^{15}N)amine
LEDER [15NH2][13C](S1)=[15N]C2=C1C=C(OC(F)(F)F)C=C2
Specifikationer
Kemiskt namn eller material Riluzole-13C,15N2
Kvantitet 1 mg
InChI formel InChI=1S/C8H5F3N2OS/c9-8(10,11)14-4-1-2-5-6(3-4)15-7(12)13-5/h1-3H,(H2,12,13)/i7+1,12+1,13+1
Formel vikt 237.0049