missing translation for 'onlineSavingsMsg'
Läs mer

Riluzole 5-Trifluoromethoxy Isomer, TRC

Produktkod. 30425704
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30425704 500 mg
30415445 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30425704 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR510015500MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 752969-85-0
Molekylformel C8H5F3N2OS
Molekylvikt (g/mol) 234.2
Synonym 5-(Trifluoromethoxy)-2-benzothiazolamine, 2-Amino-5-(trifluoromethoxy)benzothiazole, SKA 8
IUPAC-namn 5-(trifluoromethoxy)-1,3-benzothiazol-2-amine
LEDER Nc1nc2cc(OC(F)(F)F)ccc2s1

Specifications

Kemiskt namn eller material Riluzole 5-Trifluoromethoxy Isomer
Kvantitet 500 mg
InChI formel InChI=1S/C8H5F3N2OS/c9-8(10,11)14-4-1-2-6-5(3-4)13-7(12)15-6/h1-3H,(H2,12,13)
Formel vikt 234.0075
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.