Läs mer
Rimacalib, TRC
Produktkod. 30420445
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30420445 1 mg
30420442 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30420445 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR5050151MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 215174-50-8
Molekylformel C22H23FN4O2
Molekylvikt (g/mol) 394.45
Synonym SMP 114, N-[3-[(1S)-1-(2-Fluoro[1,1'-biphenyl]-4-yl)ethyl]-5-isoxazolyl]-4-morpholinecarboximidamide
IUPAC-namn N-[3-[(1S)-1-(3-fluoro-4-phenylphenyl)ethyl]-1,2-oxazol-5-yl]morpholine-4-carboximidamide
LEDER C[C@@H](c1ccc(c(F)c1)c2ccccc2)c3cc(NC(=N)N4CCOCC4)on3
Specifikationer
Kemiskt namn eller material Rimacalib
Kvantitet 1 mg
InChI formel InChI=1S/C22H23FN4O2/c1-15(17-7-8-18(19(23)13-17)16-5-3-2-4-6-16)20-14-21(29-26-20)25-22(24)27-9-11-28-12-10-27/h2-8,13-15H,9-12H2,1H3,(H2,24,25)/t15-/m0/s1
Formel vikt 394.1805