Läs mer
Tocris Bioscience™ Rimcazole dihydrochloride
GREENER_CHOICE
Produktkod. 18762514
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18762514 50 mg
18722714 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18762514 Leverantör Tocris Bioscience™ Leverantörsnummer 1497/50
σ2 antagonist. Also DAT inhibitor
Antagonist at σ receptors (IC50 values are 1480 and 386 nM at σ1 and σ2 receptors respectively). Also binds to the dopamine transporter (IC50 = 57.6 nM) and inhibits dopamine uptake. Reduces the stimulatory effects of cocaine in vivo.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 75859-03-9
Molekylformel C21H29Cl2N3
Specifikationer
Kemiskt namn eller material Rimcazole dihydrochloride
Kvantitet 50 mg
Formel vikt 394.39
Renhet >98%
Molekylvikt (g/mol) 394.384