Läs mer
Rimonabant Hydrochloride, MedChemExpress
Produktkod. 16465968
Encompass_Preferred
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
16465968 10 mg
1 options
Produktkod. 16465968 Leverantör Medchem Express Leverantörsnummer HY14137
MedChemExpress Rimonabant Hydrochloride (SR 141716A Hydrochloride) is a highly potent and selective central cannabinoid receptor (CB1) antagonist with an Ki of 1.8 nM. Rimonabant Hydrochloride (SR 141716A Hydrochloride) also inhibits Mycobacterial membrane protein Large 3 (MMPL3).
Rimonabant Hydrochloride (SR 141716A Hydrochloride) is a highly potent and selective central cannabinoid receptor (CB1) antagonist with an Ki of 1.8 nM. Rimonabant Hydrochloride (SR 141716A Hydrochloride) also inhibits Mycobacterial membrane protein Large 3 (MMPL3).
Specifikationer
Kemiskt namn eller material Rimonabant Hydrochloride
Fysisk form Solid
CAS 158681-13-1
Färg White
Kvantitet 10 mg
Anteckningar om renhetsgrad Research
Molekylformel C22H22Cl4N4O
För användning med (applikation) COVID-19-immunoregulation
Synonym SR 141716A Hydrochloride
Löslighetsinformation DMSO : 33.33 mg/mL (66.63 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
LEDER ClC1=CC=C(C2=C(C)C(C(NN3CCCCC3)=O)=NN2C4=CC=C(Cl)C=C4Cl)C=C1.Cl
Molekylvikt (g/mol) 500.25
Formel vikt 500.25
Procent renhet 99.28%