Läs mer
Rimonabant Hydrochloride, TRC
Produktkod. 18040200
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18040200 10 mg
18041320 25 mg
18030730 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18040200 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR51780010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 158681-13-1
Molekylformel C22 H21 Cl3 N4 O . Cl H
Molekylvikt (g/mol) 500.25
Synonym 1H-Pyrazole-3-carboxamide, 5-(4-chlorophenyl)-1-(2,4-dichlorophenyl)-4-methyl-N-1-piperidinyl-, hydrochloride (1:1), 1H-Pyrazole-3-carboxamide, 5-(4-chlorophenyl)-1-(2,4-dichlorophenyl)-4-methyl-N-1-piperidinyl-, monohydrochloride (9CI), 5-(4-Chlorophenyl)-1-(2,4-dichlorophenyl)-4-methyl-N-(piperidin-1-yl)-1H-pyrazole-3-carboxamide hydrochloride, Ribafit, Rimonabant hydrochloride, Riomont, SR 141716A, SR 151716A, Slimbant
IUPAC-namn 5-(4-chlorophenyl)-1-(2,4-dichlorophenyl)-4-methyl-N-piperidin-1-ylpyrazole-3-carboxamide;hydrochloride
LEDER Cl.Cc1c(nn(c2ccc(Cl)cc2Cl)c1c3ccc(Cl)cc3)C(=O)NN4CCCCC4
Specifikationer
Kemiskt namn eller material Rimonabant, Hydrochloride
Kvantitet 10 mg
InChI formel InChI=1S/C22H21Cl3N4O.ClH/c1-14-20(22(30)27-28-11-3-2-4-12-28)26-29(19-10-9-17(24)13-18(19)25)21(14)15-5-7-16(23)8-6-15;/h5-10,13H,2-4,11-12H2,1H3,(H,27,30);1H
Formel vikt 498.0548