Läs mer
Ripasudil Hydrochloride, TRC
Produktkod. 30427570
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30427570 5 mg
30419920 25 mg
30412026 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30427570 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR5179005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 934400-38-1
Molekylformel C15 H18 F N3 O2 S . 2[HCl]
Molekylvikt (g/mol) 396.3076
Synonym Isoquinoline, 4-fluoro-5-[[(2S)-hexahydro-2-methyl-1H-1,4-diazepin-1-yl]sulfonyl]-, hydrochloride (1:2), (S)-4-Fluoro-5-((2-methyl-1,4-diazepan-1-yl)sulfonyl)isoquinoline dihydrochloride, 4-Fluoro-5-[[(2S)-hexahydro-2-methyl-1H-1,4-diazepin-1-yl]sulfonyl]isoquinoline dihydrochloride, Ripasudil dihydrochloride, (2S)-1-[(4-Fluoro-5-isoquinolinyl)sulfonyl]hexahydro-2-methyl-1H-1,4-diazepine dihydrochloride
IUPAC-namn 4-fluoro-5-[[(2S)-2-methyl-1,4-diazepan-1-yl]sulfonyl]isoquinoline;hydrochloride
LEDER Cl.Cl.C[C@H]1CNCCCN1S(=O)(=O)c2cccc3cncc(F)c23
Specifikationer
Kemiskt namn eller material Ripasudil Dihydrochloride
Kvantitet 5 mg
InChI formel InChI=1S/C15H18FN3O2S.2ClH/c1-11-8-17-6-3-7-19(11)22(20,21)14-5-2-4-12-9-18-10-13(16)15(12)14;;/h2,4-5,9-11,17H,3,6-8H2,1H3;2*1H/t11-;;/m0../s1
Formel vikt 395.0637