Learn More
Invitrogen™ Human RIPK3, GST Tag Recombinant Protein
RIPK3 is a member of the receptor-interacting protein (RIP) family of serine/threonine protein kinases. Recombinant full-length human RIPK3 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
4230.00 SEK - 64700.00 SEK
Specifications
Species Human
Recombinant Recombinant
Protein Tag GST Tag
View More Specs
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15479023
View Documents Promotion Details
Invitrogen™
PV6397
10μg
4230.00 SEK
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 02-05-2024
to see stock.
15489023
View Documents Promotion Details
Invitrogen™
PV6398
100μg
14450.00 SEK
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
15499023
View Documents Promotion Details
Invitrogen™
PV6399
1mg
64700.00 SEK
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
Description
Description
RIPK3 contains a C-terminal domain unique from other RIP family members. RIPK3 is predominantly localized to the cytoplasm, and specifically, are colocalized in the mitochondrion which can undergo nucleocytoplasmic shuttling dependent on novel nuclear localization and export signals. RIPK3 is a component of the tumor necrosis factor (TNF) receptor-I signaling complex, and can induce apoptosis and weakly activate the NF-kappaB transcription factor and caspase-8 prevents RIPK3-dependent necrosis without inducing apoptosis by functioning in a proteolytically active complex with CFLAR and that this complex is required for the protective function.
  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Specifications
Specifications
Human
Recombinant
GST Tag