missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RIPK5 Recombinant Human Protein
RIPK5 is a serine/threonine kinase.
Brand: Invitrogen™ PV4679
Additional Details : Weight : 0.01000kg
Description
RIPK5 is involved in cell activation, proliferation, differentiation, and apoptosis.
Specifications
NP_056190.1 | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
1mg | |
RUO - research use only | |
RIPK5, DustyPK | |
Human | |
GST Tag | |
Frozen |
Nuclear Receptor Assay | |
132.7kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
RIPK Kinase Family | |
Recombinant | |
Baculovirus |