Läs mer
Risarestat, MedChemExpress
Produktkod. 16477048
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16477048 10 mg
16429597 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16477048 Leverantör Medchem Express Leverantörsnummer HY16433
MedChemExpress Risarestat (CT-112), an aldose reductase inhibitor, is developed for the treatment of diabetic complications.
Risarestat (CT-112), an aldose reductase inhibitor, is developed for the treatment of diabetic complications.
Specifikationer
Kemiskt namn eller material Risarestat
CAS 79714-31-1
Färg Light Yellow
Anteckningar om renhetsgrad Research
Molekylformel C16H21NO4S
Kvantitet 10 mg
Synonym CT 112
Löslighetsinformation DMSO : 105 mg/mL (324.67 mM; Need ultrasonic)
LEDER O=C(N1)SC(C2=CC=C(OCCCCC)C(OCC)=C2)C1=O
Molekylvikt (g/mol) 323.41
Formel vikt 323.41
Procent renhet 98.09%
Kvalitet Research
Fysisk form Powder
För användning med (applikation) Metabolism-sugar/lipid metabolism