Läs mer
Risperidone Isoxazole-N-oxide, TRC
Produktkod. 30179944
Change view
Kvantitet:
1 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179944 10 mg
30180332 1 mg
30179836 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179944 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR52503010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1391053-34-1
Molekylformel C23H27FN4O3
Molekylvikt (g/mol) 426.48
Synonym 3-[2-[4-(6-Fluoro-1,2-benzisoxazol-3-yl-2-oxido)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one;
IUPAC-namn 3-[2-[4-(6-fluoro-2-oxo-1,2-benzoxazol-3-yl)piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one
LEDER CC1=C(CCN2CCC(CC2)c3c4ccc(F)cc4on3=O)C(=O)N5CCCCC5=N1
Specifikationer
Kemiskt namn eller material Risperidone Isoxazole-N-oxide
Kvantitet 10 mg
InChI formel InChI=1S/C23H27FN4O3/c1-15-18(23(29)27-10-3-2-4-21(27)25-15)9-13-26-11-7-16(8-12-26)22-19-6-5-17(24)14-20(19)31-28(22)30/h5-6,14,16H,2-4,7-13H2,1H3
Formel vikt 426.2067