Läs mer
Ritanserin, MedChemExpress
Produktkod. 16456867
Encompass_Preferred
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
16456867 5 mg
1 options
Produktkod. 16456867 Leverantör Medchem Express Leverantörsnummer HY10791
MedChemExpress Ritanserin (R 55667) is a highly potent, relatively selective, orally active, long acting antagonist of 5-HT2 receptor, with an IC50 of 0.9 nM, less active on Histamine H1, Dopamine D2, Adrenergic α1, Adrenergic α2 receptors.
Ritanserin (R 55667) is a highly potent, relatively selective, orally active, long acting antagonist of 5-HT2 receptor, with an IC50 of 0.9 nM, less active on Histamine H1, Dopamine D2, Adrenergic α1, Adrenergic α2 receptors.
Specifikationer
Kemiskt namn eller material Ritanserin
CAS 87051-43-2
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C27H25F2N3OS
Kvantitet 5 mg
Synonym R 55667
Löslighetsinformation DMSO : 25 mg/mL (52.35 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
LEDER CC1=C(C(N(C=CS2)C2=N1)=O)CCN(CC/3)CCC3=C(C4=CC=C(F)C=C4)\C5=CC=C(F)C=C5
Molekylvikt (g/mol) 477.57
Formel vikt 477.57
Procent renhet 98.0%
Kvalitet Research
Fysisk form Powder
För användning med (applikation) Neuroscience-Neuromodulation