Learn More
Ritter Plastic™ Ritips™ Sterile Dispenser Tips
Used for the repetitive distribution of volumes between 0.05 and 50 mL.
6927.10 SEK - 33405.90 SEK
Specifications
Sterility | Sterile |
---|---|
For Use With (Application) | Sensitive applications, body fluid analysis |
Product Type | Pipette Tip |
For Use With (Equipment) | Wide range of pipettors |
Graduated | Yes |
Product Code | Brand | Volume (Metric) | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Quantity | Price | Quantity & Availability | ||||
16670302
|
Ritter Plastic™
40010-0008 |
0.05 mL | 100/Pk. |
33405.90 SEK
Pack of 1600 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
||||
16680302
|
Ritter Plastic™
40010-0009 |
0.5 mL | 100/Pk. |
25932.90 SEK
Pack of 1600 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
||||
16690302
|
Ritter Plastic™
40010-0010 |
1.25 mL | 100/Pk. |
25932.90 SEK
Pack of 1600 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
||||
16600312
|
Ritter Plastic™
40010-0011 |
2.5 mL | 100/Pk. |
25932.90 SEK
Pack of 1600 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
||||
16610312
|
Ritter Plastic™
40010-0012 |
5.0 mL | 100/Pk. |
25932.90 SEK
Pack of 1600 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
||||
16620312
|
Ritter Plastic™
40010-0013 |
12.5 mL | 100/Pk. |
25932.90 SEK
Pack of 1600 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
||||
16630312
|
Ritter Plastic™
40010-0014 |
25.0 mL | 25/Pk. |
6927.10 SEK
Pack of 400 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
||||
16640312
|
Ritter Plastic™
40010-0015 |
50.0 mL | 25/Pk. |
6927.10 SEK
Pack of 400 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
||||
Description
- Uses direct displacement method
- Works well for infectious and volatile liquids
- Fluid never in touch with the instrument to avoid contamination
- Automated individual tests of each single tip guarantee the tightness with water, which is documented in lot-specific records
- Black graduation marks
- Manufactured from polypropylene
- Suitable for a wide range of pipettors
- For sensitive applications or analysis of body fluids
Specifications
Sterile | |
Pipette Tip | |
Yes | |
Polypropylene |
Sensitive applications, body fluid analysis | |
Wide range of pipettors | |
RiTips |