Läs mer
Ritodrine Hydrochloride, Mikromol™
Produktkod. 17931526
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
17931526 250 mg
1 options
Produktkod. 17931526 Leverantör Mikromol Leverantörsnummer MM0800.00
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ritodrine Hydrochloride
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ritodrine Hydrochloride
CAS 23239-51-2
Analyt- eller komponentnamn Ritodrine Hydrochloride
Frakt skick Room Temperature
Molekylformel C17 H21 N O3 . HCl
Certifikat och efterlevnad ISO 17034
Synonym DU 21220, Miolene, NSC 291565, Pre-Par, Prempar, Ritodrine hydrochloride, Utemerin, Utopar, Yutopar, Benzenemethanol, 4-hydroxy-alpha-[(1R)-1-[[2-(4-hydroxyphenyl)ethyl]amino]ethyl]-, hydrochloride (1:1), (alphaS)-rel-, Benzenemethanol, 4-hydroxy-alpha-[(1R)-1-[[2-(4-hydroxyphenyl)ethyl]amino]ethyl]-, hydrochloride, (alphaS)-rel- (9CI), Benzenemethanol, 4-hydroxy-alpha-[1-[[2-(4-hydroxyphenyl)ethyl]amino]ethyl]-, hydrochloride, (R*,S*)-, Benzyl alcohol, p-hydroxy-alpha-[1-[(p-hydroxyphenethyl)amino]ethyl]-, hydrochloride, erythro- (8CI), (+/-)-Ritodrine hydrochloride
InChI formel InChI=1S/C17H21NO3.ClH/c1-12(17(21)14-4-8-16(20)9-5-14)18-11-10-13-2-6-15(19)7-3-13;/h2-9,12,17-21H,10-11H2,1H3;1H/t12-,17-;/m1./s1
LEDER Cl.C[C@@H](NCCc1ccc(O)cc1)[C@@H](O)c2ccc(O)cc2
IUPAC-namn 4-[2-[[(1S,2R)-1-hydroxy-1-(4-hydroxyphenyl)propan-2-yl]amino]ethyl]phenol;hydrochloride
Molekylvikt (g/mol) 323.81
Kvantitet 250 mg
Formel vikt 323.1288 g/mol