Läs mer
Ritodrine Hydrochloride, TRC
Produktkod. 30137148
Change view
Kvantitet:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137148 500 mg
30137333 1 g
17981855 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137148 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR533700500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 23239-51-2
Molekylformel C17H21NO3.HCl
Molekylvikt (g/mol) 323.81
Synonym Benzenemethanol, 4-hydroxy-α-[(1R)-1-[[2-(4-hydroxyphenyl)ethyl]amino]ethyl]-, hydrochloride (1:1), (αS)-rel-, Benzenemethanol, 4-hydroxy-α-[(1R)-1-[[2-(4-hydroxyphenyl)ethyl]amino]ethyl]-, hydrochloride, (αS)-rel- (9CI), Benzenemethanol, 4-hydroxy-α-[1-[[2-(4-hydroxyphenyl)ethyl]amino]ethyl]-, hydrochloride, (R*,S*)-, Benzyl alcohol, p-hydroxy-α-[1-[(p-hydroxyphenethyl)amino]ethyl]-, hydrochloride, erythro- (8CI), (±)-Ritodrine hydrochloride, DU 21220, Miolene, NSC 291565, Pre-Par, Prempar, Ritodrine hydrochloride, Utemerin, Utopar, Yutopar
IUPAC-namn 4-[2-[[(1S,2R)-1-hydroxy-1-(4-hydroxyphenyl)propan-2-yl]amino]ethyl]phenol;hydrochloride
LEDER Cl.C[C@@H](NCCc1ccc(O)cc1)[C@@H](O)c2ccc(O)cc2
Specifikationer
Kemiskt namn eller material Ritodrine Hydrochloride
Kvantitet 500 mg
InChI formel InChI=1S/C17H21NO3.ClH/c1-12(17(21)14-4-8-16(20)9-5-14)18-11-10-13-2-6-15(19)7-3-13;/h2-9,12,17-21H,10-11H2,1H3;1H/t12-,17-;/m1./s1
Formel vikt 323.1288