Läs mer
Rivastigmine N-Oxide (Technical Grade), TRC
Produktkod. 30411280
Change view
Kvantitet:
2.5 mg
Produktkod. Kvantitet
30411280 2.5 mg
1 options
Produktkod. 30411280 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR5409002.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Rivastigmine N-Oxide
CAS 1369779-37-2
Molekylformel C14 H22 N2 O3
Kvantitet 2.5 mg
Synonym N-Ethyl-N-methylcarbamic Acid 3-[(1S)-1-(Dimethylamino)ethyl]phenyl Ester N-Oxide, (S)-Ethylmethylcarbamic Acid 3-[1-(Dimethylamino)ethyl]phenyl Ester N-Oxide, [R-(R*,R*)]-2,3-Dihydroxybutanedioate N-Oxide, ENA 713 N-Oxide, Rivastigmine N-Oxide, SDZ-ENA 713 N-Oxide
InChI formel InChI=1S/C14H22N2O3/c1-6-15(3)14(17)19-13-9-7-8-12(10-13)11(2)16(4,5)18/h7-11H,6H2,1-5H3/t11-/m0/s1
LEDER CCN(C)C(=O)Oc1cccc(c1)[C@H](C)[N+](C)(C)[O-]
IUPAC-namn (1S)-1-[3-[ethyl(methyl)carbamoyl]oxyphenyl]-N,N-dimethyl-ethanamine oxide
Molekylvikt (g/mol) 266.34
Formel vikt 266.163