Läs mer
Rivastigmine N-Oxide (Technical Grade), TRC
Specifications
Specifications
| Kemiskt namn eller material | Rivastigmine N-Oxide |
| CAS | 1369779-37-2 |
| Molekylformel | C14 H22 N2 O3 |
| Kvantitet | 25 mg |
| Synonym | N-Ethyl-N-methylcarbamic Acid 3-[(1S)-1-(Dimethylamino)ethyl]phenyl Ester N-Oxide, (S)-Ethylmethylcarbamic Acid 3-[1-(Dimethylamino)ethyl]phenyl Ester N-Oxide, [R-(R*,R*)]-2,3-Dihydroxybutanedioate N-Oxide, ENA 713 N-Oxide, Rivastigmine N-Oxide, SDZ-ENA 713 N-Oxide |
| InChI formel | InChI=1S/C14H22N2O3/c1-6-15(3)14(17)19-13-9-7-8-12(10-13)11(2)16(4,5)18/h7-11H,6H2,1-5H3/t11-/m0/s1 |
| LEDER | CCN(C)C(=O)Oc1cccc(c1)[C@H](C)[N+](C)(C)[O-] |
| IUPAC-namn | (1S)-1-[3-[ethyl(methyl)carbamoyl]oxyphenyl]-N,N-dimethyl-ethanamine oxide |
| Molekylvikt (g/mol) | 266.34 |
| Formel vikt | 266.163 |