Läs mer
Rizatriptan Benzoate, TRC
Produktkod. 30180116
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180116 5 mg
30179738 50 mg
30180047 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30180116 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR5450005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 145202-66-0
Molekylformel C15 H19 N5 . C7 H6 O2
Molekylvikt (g/mol) 391.47
Synonym Rizatriptan benzoate, N,N-Dimethyl-2-[5-(1H-1,2,4-triazol-1-ylmethyl)-1H-indol-3-yl]ethanamine benzoate, 1H-Indole-3-ethanamine, N,N-dimethyl-5-(1H-1,2,4-triazol-1-ylmethyl)-, benzoate (1:1), 1H-Indole-3-ethanamine, N,N-dimethyl-5-(1H-1,2,4-triazol-1-ylmethyl)-, monobenzoate (9CI), MK 462, Maxalt, Rizora
IUPAC-namn benzoic acid;N,N-dimethyl-2-[5-(1,2,4-triazol-1-ylmethyl)-1H-indol-3-yl]ethanamine
LEDER CN(C)CCc1c[nH]c2ccc(Cn3cncn3)cc12.OC(=O)c4ccccc4
Specifikationer
Kemiskt namn eller material Rizatriptan benzoate
Kvantitet 5 mg
InChI formel InChI=1S/C15H19N5.C7H6O2/c1-19(2)6-5-13-8-17-15-4-3-12(7-14(13)15)9-20-11-16-10-18-20;8-7(9)6-4-2-1-3-5-6/h3-4,7-8,10-11,17H,5-6,9H2,1-2H3;1-5H,(H,8,9)
Formel vikt 391.2008