Läs mer
Tocris Bioscience™ RKI 1447 dihydrochloride
Beskrivning
RKI 1447 dihydrochloride is a potent and selective ROCK inhibitor (IC50 values are 6.2 and 14.5 nM for ROCK2 and ROCK1 respectively); suppresses phosphorylation of ROCK substrates MLC-2 and MYPT-1 in human cancer cells, but has no effect on the phosphorylation levels of Akt, MEK, and S6 kinase. Displays anti-invasive and antitumor activities in breast cancer cells.
Specifikationer
Specifikationer
| CAS | 1782109-09-4 |
| Kvantitet | 10 mg |
| Produkttyp | RKI 1447 dihydrochloride |
| Renhet | >99% |
| Molekylvikt (g/mol) | 399.29 |
| Inhibitorer | RKI 1447 dihydrochloride |
For Research Use Only